Invitrogen
Manufacturer Code:PA522753
Catalog # PIPA522753
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The epitope recognized by this antibody maps to a region between residues 350 and 400 of human tumor protein p53 binding protein 1 using the numbering given in entry NP&lowbar005648.1. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 53BP1; 53BP1 p202 p53-binding protein 1 p53BP1 tumor protein 53-binding protein 1 tumor protein p53-binding protein 1 tumor suppressor p53-binding protein 1; murine p53-binding protein; p53-binding protein 1; TP53-binding protein 1; transformation related protein 53 binding protein 1; transformation related protein 53 binding-protein 1; tumor protein 53-binding protein, 1; tumor protein p53-binding protein, 1; tumor suppressor p53-binding protein 1
Gene Aliases: 53BP1; m53BP1; p202; p53BP1; TDRD30; TP53; TP53BP1; Trp53bp1
UniProt ID: (Human) Q12888, (Mouse) P70399
Entrez Gene ID: (Human) 7158, (Goat) 102182833, (Rat) 296099, (Mouse) 27223
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.