Abnova
Manufacturer Code:PAB0048
Catalog # 89-136-831
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | 5-hydroxytryptophan conjugated with carrier protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C on dry atmosphere. After reconstitution with 50&muL of distilled water and 50&muL of glycerol,store at -20°C for 2 years. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against 5-hydroxytryptophan.
For Research Use Only
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.