Novus Biologicals
Manufacturer Code:NBP270166V
Catalog # NBP270166V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3F1 |
Immunogen | Recombinant protein fragment corresponding to amino acids 18-364 of human 5-Lipoxygenase (NP_000689) produced in E.coli. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 5-lipoxygenase 5-LO 5-LOX 5LPG arachidonate 5-lipoxygenase arachidonic 5-lipoxygenase alpha-10 isoform arachidonic 5-lipoxygenase delta-10-13 isoform arachidonic 5-lipoxygenase delta-13 isoform arachidonic 5-lipoxygenase delta-p10 isoform arachidonic acid 5-lipoxygenase EC 1.13.11 EC 1.13.11.34 leukotriene A4 synthase LOG5 MGC163204; 5-LO; Arachidonate 5-lipoxygenase; arachidonic 5-lipoxygenase alpha-10 isoform; arachidonic 5-lipoxygenase delta-10-13 isoform; arachidonic 5-lipoxygenase delta-13 isoform; arachidonic 5-lipoxygenase delta-p10 isoform; arachidonic acid 5-lipoxygenase; leukotriene A4 synthase; LOX-5; Polyunsaturated fatty acid 5-lipoxygenase
Gene Aliases: 5-LO; 5-LOX; 5LPG; ALOX5; LOG5
UniProt ID: (Human) P09917
Entrez Gene ID: (Human) 240
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.