R&D Systems
Manufacturer Code:FAB10325S100UG
Catalog # FAB10325S100U
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1015322 |
Immunogen | Synthetic peptide containing human 5-HTR7 Accession &num P34969 |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 5-HT-7; 5-HT-X; 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled); 5-hydroxytryptamine (serotonin) receptor 7, adenylate cyclase-coupled; 5-hydroxytryptamine receptor 7; 5HT7 5-HT-7 5-HT75-hydroxytryptamine receptor 7 5-HT-X 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled) HTR7 serotonin 5-HT-7 receptor Serotonin receptor 7; Serotonin receptor 7
Gene Aliases: 5-HT7; HTR7
UniProt ID: (Human) P34969
Entrez Gene ID: (Human) 3363
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.