Novus Biologicals
Manufacturer Code:NBP2825400.1ML
Catalog # NB030469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human 5-HT3E |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT3 receptor subunit E splice variant HTR3Ea 5-HT3c15-HT3E 5-hydroxytryptamine (serotonin) receptor 3 family member E 5-hydroxytryptamine receptor 3E MGC120035 MGC120036 serotonin receptor 3E; 5-HT3-E; 5-hydroxytryptamine (serotonin) receptor 3, family member E; 5-hydroxytryptamine (serotonin) receptor 3E, ionotropic; 5-hydroxytryptamine receptor 3 subunit E; 5-hydroxytryptamine receptor 3E; Serotonin receptor 3E
Gene Aliases: 5-HT3-E; 5-HT3c1; 5-HT3E; HTR3E
UniProt ID: (Human) A5X5Y0
Entrez Gene ID: (Human) 285242
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.