Novus Biologicals
Manufacturer Code:NBP151969
Catalog # NBP151969
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-SNYLQTQDQTDQQE corresponding to internal region (near C-terminus) according to NP_006019.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT3-B; 5-HT3-B 5-HT3B5-hydroxytryptamine receptor 3B 5-hydroxytryptamine (serotonin) receptor 3B 5-hydroxytryptamine 3 receptor B subunit Serotonin receptor 3B serotonin-gated ion channel subunit; 5-hydroxytryptamine (serotonin) receptor 3B, ionotropic; 5-hydroxytryptamine 3 receptor B subunit; 5-hydroxytryptamine receptor 3B; Serotonin receptor 3B; serotonin-gated ion channel subunit
Gene Aliases: 5-HT3B; HTR3B
UniProt ID: (Human) O95264
Entrez Gene ID: (Human) 9177
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.