Novus Biologicals
Manufacturer Code:NB10041382
Catalog # NB10041382
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-GPQDFEKSPRDR corresponding to internal region according to NP_998786.2 NP_000860.2 NP_001155244.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT-3; 5-HT-35-HT3-A 5-HT3R5-hydroxytryptamine receptor 3A 5-hydroxytryptamine (serotonin) receptor 3A 5-hydroxytryptamine receptor 35-HT3A5HT3R HTR35HT3 serotonin receptor Serotonin receptor 3A Serotonin-gated ion channel receptor; 5-HT3-A; 5-hydroxytryptamine (serotonin) receptor 3A, ionotropic; 5-hydroxytryptamine receptor 3; 5-hydroxytryptamine receptor 3A; 5HT3 serotonin receptor; Serotonin receptor 3A; Serotonin-gated ion channel receptor
Gene Aliases: 5-HT-3; 5-HT3A; 5-HT3R; 5HT3R; HTR3; HTR3A
UniProt ID: (Human) P46098
Entrez Gene ID: (Human) 3359
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.