Novus Biologicals
Manufacturer Code:NB1001524
Catalog # NB1001524
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-QVENLELPVN corresponding to internal region according to NP_000859.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT-1C; 5-HT-2C; 5-HT1C 5-HT-1C 5-HT-2C 5-HT2C5-hydroxytryptamine receptor 1C 5-HTR2C 5-hydroxytryptamine (serotonin) receptor 2C 5-hydroxytryptamine receptor 2C HTR1C serotonin 5-HT-2C receptor Serotonin receptor 2C; 5-hydroxytryptamine (serotonin) receptor 2C, G protein-coupled; 5-hydroxytryptamine receptor 1C; 5-hydroxytryptamine receptor 2C; serotonin 5-HT-1C receptor; serotonin 5-HT-2C receptor; Serotonin receptor 2C
Gene Aliases: 5-HT1C; 5-HT2C; 5-HTR2C; 5HTR2C; HTR1C; HTR2C
UniProt ID: (Human) P28335
Entrez Gene ID: (Human) 3358
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.