R&D Systems
Manufacturer Code:MAB10322100
Catalog # MAB10322100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1014838 |
Immunogen | Synthetic peptide containing human 5- HTR2B Accession &num P41595 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 5-HT 2B receptor; 5-HT(2B) 5HT2B 5-HT-2B 5-HT2B5-HT 2B receptor 5-hydroxytryptamine (serotonin) receptor 2B 5-hydroxytryptamine 2B receptor 5-hydroxytryptamine receptor 2B HTR2B Serotonin receptor 2B; 5-HT-2B; 5-hydroxytryptamine (serotonin) receptor 2B, G protein-coupled; 5-hydroxytryptamine 2B receptor; 5-hydroxytryptamine receptor 2B; 5-hydroxytryptamine receptor 2B variant b; Serotonin receptor 2B
Gene Aliases: 5-HT(2B); 5-HT-2B; 5-HT2B; HTR2B
UniProt ID: (Human) P41595
Entrez Gene ID: (Human) 3357
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.