Novus Biologicals
Manufacturer Code:NBP278806
Catalog # NB002959
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | This 5-HT2A antibody is made to a peptide with sequence KNSKQDAKTTDNDCS corresponding to internal region according to NP_000612.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
Protein Aliases: 5-HT-2; 5-HT-2 5-HT2 receptor 5-HT-2A 5-hydroxytryptamine (serotonin) receptor 2A 5-hydroxytryptamine receptor 2A HTR25-HT2A serotonin 5-HT-2A receptor Serotonin receptor 2A; 5-HT2 receptor; 5-hydroxytryptamine (serotonin) receptor 2A, G protein-coupled; 5-hydroxytryptamine receptor 2A; serotonin 5-HT-2A receptor; Serotonin receptor 2A
Gene Aliases: 5-HT2A; HTR2; HTR2A
UniProt ID: (Human) P28223
Entrez Gene ID: (Human) 3356
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.