Novus Biologicals
Manufacturer Code:NB10056349
Catalog # NB10056349
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was generated by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 1-18 and 251-267 of rat 5-HT1DR (Genbank accession no. NP_036984). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT-1D; 5-HT-1D 5-HT-1D-alpha 5-HT1DHTR1DA 5-hydroxytryptamine (serotonin) receptor 1D HT1DA HTRL RDC45-hydroxytryptamine receptor 1D Serotonin receptor 1D; 5-HT-1D-alpha; 5-hydroxytryptamine (serotonin) receptor 1D, G protein-coupled; 5-hydroxytryptamine receptor 1D; Serotonin 1D alpha receptor; Serotonin receptor 1D
Gene Aliases: 5-HT1D; HT1DA; HTR1D; HTR1DA; HTRL; RDC4
UniProt ID: (Human) P28221
Entrez Gene ID: (Human) 3352
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.