R&D Systems
Manufacturer Code:MAB5795
Catalog # MAB5795
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 606112 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human 5'-Nucleotidase/CD73 Trp27-Ile511 Accession # P21589 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 5' nucleotidase (CD73) 5'-NT 5-NT 5'Nucleotidase 5'-nucleotidase ecto (CD73) CD73 CD73 antigen E5NT EC 3.1.3.5 ecto-5'-nucleotidase eN eNT NT NT55'-nucleotidase NT5E NTE Purine 5-Prime-Nucleotidase; 5'-NT; 5'-nucleotidase; 5'-nucleotidase, ecto (CD73); CD73; Ecto-5'-nucleotidase; Purine 5-Prime-Nucleotidase
Gene Aliases: CALJA; CD73; E5NT; eN; eNT; NT; NT5; NT5E; NTE
UniProt ID: (Human) P21589
Entrez Gene ID: (Human) 4907
Molecular Function:
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.