Novus Biologicals
Manufacturer Code:NBP292960
Catalog # NB027757
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 50 to the C-terminus of human EIF4EBP1 (NP_004086.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4E-BP1; 4E-BP1 eIF4E-binding protein 1 eukaryotic translation initiation factor 4E binding protein 14EBP14E-BP1BP-1 eukaryotic translation initiation factor 4E-binding protein 1 PHAS-IMGC4316 Phosphorylated heat- and acid-stable protein regulated by insulin 1; eIF4E-binding protein 1; Eukaryotic translation initiation factor 4E-binding protein 1; PHAS-I; Phosphorylated heat- and acid-stable protein regulated by insulin 1
Gene Aliases: 4E-BP1; 4EBP1; BP-1; EIF4EBP1; PHAS-I
UniProt ID: (Human) Q13541
Entrez Gene ID: (Human) 1978
Molecular Function:
RNA binding protein
nucleic acid binding
translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.