Invitrogen
Manufacturer Code:PA561027
Catalog # PIPA561027
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human 4E-BP3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4E-BP3; eIF4E-binding protein 3; eukaryotic initiation factor 4E-binding protein 3; Eukaryotic translation initiation factor 4E-binding protein 3; NG36
Gene Aliases: 4E-BP3; 4EBP3; EIF4EBP3
UniProt ID: (Human) O60516
Entrez Gene ID: (Human) 8637
Molecular Function:
RNA binding protein
nucleic acid binding
translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.