Invitrogen
Manufacturer Code:PA589361
Catalog # PIPA589361
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 61-120 of human EIF4EBP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4E-BP2; eIF4E-binding protein 2; Eukaryotic translation initiation factor 4E-binding protein 2; phosphorylated, heat and acid stable regulated by insulin protein II
Gene Aliases: 4EBP2; EIF4EBP2; PHASII
UniProt ID: (Human) Q13542
Entrez Gene ID: (Human) 1979
Molecular Function: RNA binding protein nucleic acid binding translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.