Invitrogen
Manufacturer Code:PA557941
Catalog # PIPA557941
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human 4E-BP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4E-BP2; 4E-BP2 eIF4E-binding protein 2 eukaryotic translation initiation factor 4E-binding protein 2 phosphorylated heat and acid stable regulated by insulin protein II; eIF4E-binding protein 2; Eukaryotic translation initiation factor 4E-binding protein 2; phosphorylated, heat and acid stable regulated by insulin protein II
Gene Aliases: 4EBP2; EIF4EBP2; PHASII
UniProt ID: (Human) Q13542
Entrez Gene ID: (Human) 1979
Molecular Function: RNA binding protein nucleic acid binding translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.