Invitrogen
Manufacturer Code:MA527577
Catalog # PIMA527577
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 12F4 |
Immunogen | Synthetic 4-Hydroxy-2-hexenal modified Keyhole Limpet Hemocyanin (KLH) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 4-Hydroxy-2-hexenal (4-HHE) 4-Hydroxy-2-hexenal 4-HHE MG MG-modified protein 4-Hydroxy-2-hexenal-modified 4-Hydroxy-2-hexenal 4-hydroxy Hexenal HHE 4-HHE Hydroxyhexenal
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.