Novus Biologicals
Manufacturer Code:NBP246164
Catalog # NBP246164
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5H5 |
Immunogen | Human recombinant protein fragment corresponding to amino acids 24-186 of human TNFRSF9 (NP_001552) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 4-1BB 4-1BB ligand receptor CD137 antigen CD137MGC2172 CDw137 FLJ43501 ILAhomolog of mouse 4-1BB induced by lymphocyte activation (ILA) interleukin-activated receptor homolog of mouse Ly63 receptor protein 4-1BB T cell antigen ILA T-cell antigen 4-1BB homolog T-cell antigen ILA tumor necrosis factor receptor superfamily member 9 tumor necrosis factor receptor superfamily member 9; 4-1BB ligand receptor; CD137; CD137 antigen; CDw137; homolog of mouse 4-1BB; induced by lymphocyte activation (ILA); interleukin-activated receptor, homolog of mouse Ly63; receptor protein 4-1BB; T cell antigen ILA; T-cell antigen 4-1BB homolog; T-cell antigen ILA; Tumor necrosis factor receptor superfamily member 9; tumor necrosis factor receptor superfamily, member 9
Gene Aliases: 4-1BB; CD137; CDw137; ILA; TNFRSF9
UniProt ID: (Human) Q07011
Entrez Gene ID: (Human) 3604
Molecular Function:
cytokine receptor
receptor
tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.