Novus Biologicals
Manufacturer Code:NBP290388100UL
Catalog # NB022229
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 14 |
Immunogen | Recombinant Mouse 4-1BB Ligand/TNFSF9 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 4-1BB ligand; 4-1BB ligand 4-1BBL 4-1BB-L CD137L homolog of mouse 4-1BB-L receptor 4-1BB ligand tumor necrosis factor (ligand) superfamily member 9 tumor necrosis factor ligand superfamily member 9; 4-1BBL; homolog of mouse 4-1BB-L; receptor 4-1BB ligand; tumor necrosis factor (ligand) superfamily member 9; tumor necrosis factor (ligand) superfamily, member 9; tumor necrosis factor ligand 5A; Tumor necrosis factor ligand superfamily member 9
Gene Aliases: 4-1BB-L; CD137L; TNFSF9; TNLG5A
UniProt ID: (Human) P41273
Entrez Gene ID: (Human) 8744
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.