Abnova
Manufacturer Code:H00000622B01P
Catalog # 89-016-729
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | BDH1 (NP_004042.1 1 a.a. ~ 343 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human BDH1 protein.
For Research Use Only
Protein Aliases: (R)-3-hydroxybutyrate dehydrogenase; 3-hydroxybutyrate dehydrogenase; 3-hydroxybutyrate dehydrogenase (heart, mitochondrial); BDH; D-beta-hydroxybutyrate dehydrogenase, mitochondrial; MGC2723; MGC4347; MGC9788; SDR9C1; Short chain dehydrogenase/reductase family 9C member 1; short chain dehydrogenase/reductase family 9C, member 1
Gene Aliases: BDH; BDH1; SDR9C1
UniProt ID: (Human) Q02338
Entrez Gene ID: (Human) 622
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.