R&D Systems
Manufacturer Code:MAB10393SP
Catalog # MAB10393SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Blocking Assay (BLOCK) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 999602 |
Immunogen | NS0 mouse myeloma cell line transfected with human 2B4/CD244/SLAMF4 Gly19-His222 Accession &num NP_057466 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD244; CD244 CD244 antigen CD244 molecule natural killer cell receptor 2B4 h2B4 NAIL NAILNK cell activation-inducing ligand natural killer cell receptor 2B4 NKR2B4 NKR2B4NK cell type I receptor protein 2B42B4CD244 natural killer cell receptor 2B4 Nmrk SLAMF4 SLAMF4NK cell activation inducing ligand NAIL; CD244 molecule, natural killer cell receptor 2B4; h2B4; NAIL; Natural killer cell receptor 2B4; NK cell activation inducing ligand NAIL; NK cell activation-inducing ligand; NK cell type I receptor protein 2B4; NKR2B4; Signaling lymphocytic activation molecule 4; SLAM family member 4; SLAMF4
Gene Aliases: 2B4; CD244; NAIL; NKR2B4; Nmrk; SLAMF4
UniProt ID: (Human) Q9BZW8
Entrez Gene ID: (Human) 51744
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein immunoglobulin receptor superfamily kinase protein kinase receptor signaling molecule transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.