Proteintech
Manufacturer Code:188301AP150UL
Catalog # 50-173-2247
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: down regulated in metastasis; Down-regulated in metastasis protein; DRIM NNP73 Novel nucleolar protein 73 Protein Key 1A6 UTP20; NNP73; Novel nucleolar protein 73; Protein Key-1A6; Small subunit processome component 20 homolog; UTP20 small subunit (SSU) processome component; UTP20 small subunit processome component; UTP20, small subunit (SSU) processome component, homolog
Gene Aliases: 1A6/DRIM; 3830408P06Rik; AA617408; DRIM; mDRIM; mDRIN; RGD1560606; UTP20
UniProt ID: (Human) O75691
Entrez Gene ID: (Human) 27340, (Mouse) 70683, (Rat) 314713
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.