R&D Systems
Manufacturer Code:MAB7515
Catalog # MAB7515
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 736043 |
Immunogen | <EM>E. coli</EM>-derived recombinant human 17 beta-HSD14/HSD17B14 Ala2-Ser270 Accession # Q9BPX1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 17 betaHSD14 dehydrogenase/reductase (SDR family) member 10 Dehydrogenase/reductase SDR family member 10 DHRS10 DHRS1017-beta-hydroxysteroid dehydrogenase 14 EC 1.1.1.- HSD17B14 hydroxysteroid (17-beta) dehydrogenase 1417-beta-hydroxysteroid dehydrogenase DHRS10 retinal short-chain dehydrogenase/reductase 3 Retinal short-chain dehydrogenase/reductase retSDR3 retSDR3 RETSDR317-beta-HSD 14 SDR3 SDR47C1 short chain dehydrogenase/reductase family 47C member 1; 17-beta-HSD 14; 17-beta-hydroxysteroid dehydrogenase 14; 17-beta-hydroxysteroid dehydrogenase DHRS10; dehydrogenase/reductase (SDR family) member 10; Dehydrogenase/reductase SDR family member 10; hydroxysteroid (17-beta) dehydrogenase 14; retinal short-chain dehydrogenase/reductase 3; Retinal short-chain dehydrogenase/reductase retSDR3; Short chain dehydrogenase/reductase family 47C member 1; short chain dehydrogenase/reductase family 47C, member 1; testicular tissue protein Li 52
Gene Aliases: DHRS10; HSD17B14; retSDR3; SDR3; SDR47C1; UNQ502/PRO474
UniProt ID: (Human) Q9BPX1
Entrez Gene ID: (Human) 51171
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.