Novus Biologicals
Manufacturer Code:NBP296996AF532
Catalog # NB080581
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant 17 beta-HSD14/HSD17B14 (NP_057330.2 Met1-Ser270). |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 17-beta-HSD 14; 17-beta-hydroxysteroid dehydrogenase 14; 17-beta-hydroxysteroid dehydrogenase DHRS10; dehydrogenase/reductase (SDR family) member 10; dehydrogenase/reductase (SDR family) member 10 Dehydrogenase/reductase SDR family member 10 DHRS1017-beta-hydroxysteroid dehydrogenase 14 EC 1.1.1.- hydroxysteroid (17-beta) dehydrogenase 1417-beta-hydroxysteroid dehydrogenase DHRS10 retinal short-chain dehydrogenase/reductase 3 Retinal short-chain dehydrogenase/reductase retSDR3 retSDR3 RETSDR317-beta-HSD 14 SDR3 SDR47C1 short chain dehydrogenase/reductase family 47C member 1; Dehydrogenase/reductase SDR family member 10; hydroxysteroid (17-beta) dehydrogenase 14; retinal short-chain dehydrogenase/reductase 3; Retinal short-chain dehydrogenase/reductase retSDR3; Short chain dehydrogenase/reductase family 47C member 1; short chain dehydrogenase/reductase family 47C, member 1; testicular tissue protein Li 52
Gene Aliases: DHRS10; HSD17B14; retSDR3; SDR3; SDR47C1; UNQ502/PRO474
UniProt ID: (Human) Q9BPX1
Entrez Gene ID: (Human) 51171
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.