Invitrogen
Manufacturer Code:PA581402
Catalog # PIPA581402
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Mouse HPGD/15-PGDH protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 15-hydroxyprostaglandin dehydrogenase; 15-hydroxyprostaglandin dehydrogenase &lsqbNAD+] Prostaglandin dehydrogenase 1 HPGD PGDH1; 15-hydroxyprostaglandin dehydrogenase [NAD(+)]; 15-PGDH; PGDH; Prostaglandin dehydrogenase 1
Gene Aliases: 15-PGDH; AV026552; Hpgd; Pgdh1
UniProt ID: (Mouse) Q8VCC1
Entrez Gene ID: (Mouse) 15446
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.