Novus Biologicals
Manufacturer Code:NBP290287AF594
Catalog # NB071872
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 27 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human 15-Lipoxygenase 2 (AAH35217.1 Met1-Ile676). |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 15-lipoxygenase 2; 15-lipoxygenase 2 15-LOX-B arachidonate 15-lipoxygenase 2 arachidonate 15-lipoxygenase B Arachidonate 15-lipoxygenase type II arachidonate 15-lipoxygenase second type arachidonate 15-lipoxygenase type B arachidonate omega(6) lipoxygenase EC 1.13.11 EC 1.13.11.3315-LOX-215S-lipoxygenase; 15-LOX-2; 15-LOX-B; 15S-lipoxygenase; arachidonate 15-lipoxygenase 2; Arachidonate 15-lipoxygenase B; Arachidonate 15-lipoxygenase type II; arachidonate 15-lipoxygenase, second type; arachidonate omega(6) lipoxygenase; Linoleate 13-lipoxygenase 15-LOb; Polyunsaturated fatty acid lipoxygenase ALOX15B
Gene Aliases: 15-LOX-2; ALOX15B
UniProt ID: (Human) O15296
Entrez Gene ID: (Human) 247
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.