Invitrogen
Manufacturer Code:PA527641
Catalog # PIPA527641
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 245 of Human 14-3-3 theta |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 14-3-3 1C5 HS1 14-3-3 protein T-cell 14-3-3 protein tau 14-3-3 protein theta protein tau; 14-3-3 protein T-cell; 14-3-3 protein tau; 14-3-3 protein theta; 14-3-3 tau; 14-3-3 theta; Protein HS1; protein, theta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, theta polypeptide
Gene Aliases: 14-3-3; 1C5; 2700028P07Rik; AA409740; AU021156; HS1; R74690; YWHAQ
UniProt ID: (Human) P27348, (Mouse) P68254
Entrez Gene ID: (Human) 10971, (Mouse) 22630
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.