Invitrogen
Manufacturer Code:MA535997
Catalog # PIMA535997
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 02 |
Immunogen | Recombinant Human 14-3-3 sigma/Stratifin/YWHAS Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 14-3-3 protein sigma; 14-3-3 protein sigma 14-3-3 sigma 14-3-3 sigma protein 14-3-3sigma BOS_2359 Epithelial cell marker protein 1 Er HGNC:10773 HME1 Mkrn3 Mme1 SFN Stratifin tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein sigma polypeptide YWHAS; 14-3-3 sigma; Epithelial cell marker protein 1; Stratifin
Gene Aliases: HME1; SFN; YWHAS
UniProt ID: (Human) P31947
Entrez Gene ID: (Human) 2810
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.