Invitrogen
Manufacturer Code:PA586412
Catalog # PIPA586412
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 40-90 of Human 14-3-3 sigma. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 14-3-3 protein sigma; 14-3-3 protein sigma Epithelial cell marker protein 1 Stratifin SFN HME1 YWHAS; 14-3-3 sigma; 14-3-3 sigma protein; Epithelial cell marker protein 1; Stratifin; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, sigma polypeptide
Gene Aliases: Er; HME1; Mkrn3; Mme1; SFN; YWHAS
UniProt ID: (Human) P31947, (Mouse) O70456
Entrez Gene ID: (Human) 2810, (Mouse) 55948
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.