Invitrogen
Manufacturer Code:PA523507
Catalog # PIPA523507
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (133 EVATGDDKKRIIDSARSA 150) of 14-3-3 sigma protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 14-3-3 protein sigma; 14-3-3 sigma; Epithelial cell marker protein 1; Stratifin; YWHAS 14-3-3 protein sigma 14-3-3 sigma epithelial cell marker protein 1
Gene Aliases: HME1; SFN; YWHAS
UniProt ID: (Human) P31947, (Bovine) Q0VC36
Entrez Gene ID: (Human) 2810, (Bovine) 528453, (Horse) 100057082, (Dog) 487351
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.