Invitrogen
Manufacturer Code:PA518462
Catalog # PIPA518462
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (DLHTLSEDSYKDST) corresponding to the internal amino acids of SFN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 14-3-3 protein sigma; 14-3-3 sigma; Epithelial cell marker protein 1; SFN 14-3-3sigma stratifin HGNC:10773 YWHAS; Stratifin
Gene Aliases: HME1; SFN; YWHAS
UniProt ID: (Human) P31947
Entrez Gene ID: (Human) 2810
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.