Novus Biologicals
Manufacturer Code:NB100406SS
Catalog # NB100406SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | KC21 |
Immunogen | N-terminal fragment of human 14-3-3 gamma where the N-terminal Val was acetylated [UniProt# P61981] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 14-3-3 gamma; 14-3-3 gamma 14-3-3 protein gamma 14-3-3GAMMA KCIP-1 Protein kinase C inhibitor protein 1 tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gammapolypeptide; 14-3-3 protein gamma; 14-3-3 protein gamma, N-terminally processed; KCIP-1; Protein kinase C inhibitor protein 1; protein phosphatase 1, regulatory subunit 170; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide
Gene Aliases: 14-3-3GAMMA; PPP1R170; YWHAG
UniProt ID: (Human) P61981
Entrez Gene ID: (Human) 7532
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.