Novus Biologicals
Manufacturer Code:NBP250579G
Catalog # NBP250579G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AT4B9 |
Immunogen | Recombinant human 14-3-3 gamma (1-247aa) purified from E. coli |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 14-3-3 gamma; 14-3-3 gamma 14-3-3 protein gamma 14-3-3GAMMA KCIP-1 Protein kinase C inhibitor protein 1 tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gammapolypeptide; 14-3-3 protein gamma; 14-3-3 protein gamma, N-terminally processed; KCIP-1; Protein kinase C inhibitor protein 1; protein phosphatase 1, regulatory subunit 170; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide
Gene Aliases: 14-3-3GAMMA; PPP1R170; YWHAG
UniProt ID: (Human) P61981
Entrez Gene ID: (Human) 7532
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.