Invitrogen
Manufacturer Code:MA535769
Catalog # PIMA535769
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1457 |
Immunogen | A synthesized peptide derived from human 14-3-3 gamma |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 14-3-3 gamma; 14-3-3 protein gamma; 14-3-3 protein gamma 14-3-3 protein gamma N-terminally processed 14-3-3 protein gamma-1 14-3-3G 14-3-3gamma BOS_22941 cb47 D7Bwg1348e fd20c02 KCIP-1 PPP1R170 Protein kinase C inhibitor protein 1 RCJMB04_5e12 SRCRB4D wu:fd20c02 wu:fj53d01 Ywhag ywhag1 zgc:73131; 14-3-3 protein gamma subtype; 14-3-3 protein gamma, N-terminally processed; 14-3-3 protein gamma-subtype; 3-monooxgenase/tryptophan 5-monooxgenase activation protein, gamma polypeptide; 3-monooxgenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide; 3-monooxygenase/tryptophan 5-monooxgenase activation protein gamma polypeptide; 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide; KCIP-1; Protein kinase C inhibitor protein 1; protein phosphatase 1, regulatory subunit 170; tryosine 3-monooxgenase/tryptophan 5 monooxgenase activation protein gamma; tryosine 3-monooxgenase/tryptophan 5-monooxgenase activation protein gamma polypeptide; tyrosine 3-monooxgenase/tryptophan 5-monooxgenase activation protein, gamma polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein gamma polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, gamma polypeptide
Gene Aliases: 14-3-3G; 14-3-3GAMMA; D7Bwg1348e; PPP1R170; YWHAG
UniProt ID: (Human) P61981, (Rat) P61983, (Mouse) P61982
Entrez Gene ID: (Human) 7532, (Rat) 56010, (Mouse) 22628
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.