Invitrogen
Manufacturer Code:PA5116415
Catalog # PIPA5116415
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 137-162 amino acids from the Central region of DANRE ywhag1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 14-3-3 gamma 14-3-3 protein gamma 14-3-3 protein gamma subtype 14-3-3 protein gamma N-terminally processed 14-3-3 protein gamma-1 14-3-3 protein gamma-subtype 14-3-3G 14-3-3gamma 3-monooxgenase/tryptophan 5-monooxgenase activation protein gamma polypeptide 3-monooxgenase/tryptophan 5-monooxygenase activation protein gamma polypeptide 3-monooxygenase/tryptophan 5-monooxgenase activation protein gamma polypeptide 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma polypeptide 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma polypeptide 1 cb47 D7Bwg1348e fd20c02 KCIP-1 PPP1R170 protein kinase C inhibitor protein 1 protein phosphatase 1 regulatory subunit 170 RCJMB04_5e12 scavenger receptor cysteine rich domain containing group B (4 domains) SRCRB4D tryosine 3-monooxgenase/tryptophan 5 monooxgenase activation protein gamma tryosine 3-monooxgenase/tryptophan 5-monooxgenase activation protein gamma polypeptide tyrosine 3/tryptophan 5 -monooxygenase activation protein gamma polypeptide tyrosine 3/tryptophan 5 -monooxygenase activation protein gamma polypeptide tyrosine 3-monooxgenase/tryptophan 5-monooxgenase activation protein gamma polypeptide tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma polypeptide wu:fd20c02 wu:fj53d01 ywhag ywhag1 zgc:73131; 14-3-3 protein gamma-1
Gene Aliases: cb47; fd20c02; wu:fd20c02; wu:fj53d01; ywhag; ywhag1; zgc:73131
UniProt ID: (Zebrafish) Q6PC29
Entrez Gene ID: (Zebrafish) 117604
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.