Invitrogen
Manufacturer Code:PA186556
Catalog # PIPA186556
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide of acetylated N-terminal amino acids 1-10 of sheep. Peptide sequence: Ac. V D R E Q L V Q K A C of sheep 14-3-3 gamma. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C or Store at -20°C if preferred |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 14-3-3 gamma; 14-3-3 protein gamma; 14-3-3 protein gamma, N-terminally processed; 14-3-3gamma D7Bwg1348e; KCIP-1; Protein kinase C inhibitor protein 1; protein phosphatase 1, regulatory subunit 170; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide
Gene Aliases: 14-3-3GAMMA; PPP1R170; YWHAG
UniProt ID: (Human) P61981
Entrez Gene ID: (Human) 7532
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.