Invitrogen
Manufacturer Code:MA529648
Catalog # PIMA529648
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6 |
Immunogen | Recombit Human 14-3-3 eta/YWHAH protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 14-3-3 eta; 14-3-3 protein eta; Protein AS1; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, eta polypeptide; YWHA1 14-3-3 eta 14-3-3 protein eta
Gene Aliases: YWHA1; YWHAH
UniProt ID: (Human) Q04917
Entrez Gene ID: (Human) 7533
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.