Novus Biologicals
Manufacturer Code:NBP192691IR
Catalog # NBP192691IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3G12 |
Immunogen | Full length recombinant human 14-3-3 eta expressed in and purified from E.coli [UniProt# Q04917]. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 14-3-3 eta; 14-3-3 protein eta; 14-3-3 protein eta Protein AS1 tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein etapolypeptide YWHA114-3-3 eta; Protein AS1; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, eta polypeptide
Gene Aliases: YWHA1; YWHAH
UniProt ID: (Human) Q04917
Entrez Gene ID: (Human) 7533
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.