Invitrogen
Manufacturer Code:PA575298
Catalog # PIPA575298
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 50-100 of Human 14-3-3 eta. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 14-3-3 eta; 14-3-3 protein eta; 14-3-3 protein eta Protein AS1 YWHAH YWHA1 1433F; 14-3-3 protein eta-subtype; Protein AS1; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, eta polypeptide
Gene Aliases: 14-3-3e; YWHA1; YWHAH
UniProt ID: (Human) Q04917, (Rat) P68511, (Mouse) P68510
Entrez Gene ID: (Human) 7533, (Rat) 25576, (Mouse) 22629
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.