R&D Systems
Manufacturer Code:AF4420SP
Catalog # AF4420SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human 14-3-3 eta Met1-Asn246 Accession # Q04917 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 14-3-3 eta; 14-3-3 protein eta; 1433 eta 14-3-3 protein eta Protein AS1 tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein etapolypeptide YWHA1 YWHA114-3-3 eta YWHAH; Protein AS1; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, eta polypeptide
Gene Aliases: YWHA1; YWHAH
UniProt ID: (Human) Q04917
Entrez Gene ID: (Human) 7533
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.