Novus Biologicals
Manufacturer Code:NBP267831
Catalog # NBP267831
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SD084-02 |
Immunogen | Full length protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: 14-3-3 protein T-cell; 14-3-3 protein tau; 14-3-3 protein tau 14-3-3 protein T-cell 14-3-3 protein theta 14-3-31C5 HS1 Protein HS1 protein tau tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein thetapolypeptide; 14-3-3 protein theta; Protein HS1; protein, theta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, theta polypeptide
Gene Aliases: 14-3-3; 1C5; HS1; YWHAQ
UniProt ID: (Human) P27348
Entrez Gene ID: (Human) 10971
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.