Invitrogen
Manufacturer Code:PA570131
Catalog # PIPA570131
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of mouse 1190002N15Rik |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deleted in autism protein 1 homolog; Divergent protein kinase domain 2A; Golgi protein GoPro49; Golgi Protein of 49 kDa; GoPro49; HASF; Hypoxia and AKT-induced stem cell factor; UPF0672 protein C3orf58 homolog
Gene Aliases: Dipk2a; GoPro49; HASF
UniProt ID: (Mouse) Q3USZ8
Entrez Gene ID: (Mouse) 68861
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.