Abnova
Manufacturer Code:PAB29156
Catalog # 89-130-611
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to internal region of <i>Arabidopsis thaliana</i> SEUSS emulsified in Freund's adjuvant. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Chicken polyclonal antibody raised against Arabidopsis thaliana SEUSS.
For Research Use Only
Protein Aliases: AtSEU; F28H19.10; F28H19_10; SEUSS; Transcriptional corepressor SEUSS
Gene Aliases: AT1G43850; F28H19.10; F28H19_10; SEU; seuss
UniProt ID: (Arabidopsis) Q8W234
Entrez Gene ID: (Arabidopsis) 840981
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.