Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Uric Acid Liquid Reagents
Reagent requires Uric Acid Standard (23-666-424)/Chemistry Calibrator (23-666-189)
Supplier: Pointe Scientific HU982615
This item is not returnable.
View return policy
Description
Method: Uricase/trinder, endpoint
Storage Temperature: 2–8°C
Reconstituted Stability: Ready-to-use liquid
Linearity: 20mg/dL
Measurement Wavelength: 520nm
Reaction Temperature: 37°C
Expected Values: 2.5–7.7mg/dL
Specifications
CE Marked | |
4-AAP >0.2mM, HDCBS 2mM, Uricase (microbial) >150U/L, Peroxidase (horseradish) >2500U/L, pH 7.5 ± 0.1 Buffer, non-reactive Stabilizers | |
8.1 | |
Liquid |
Uric Acid Reagent Set | |
Quantitative determination of Uric Acid in Serum, in-vitro diagnostic use only | |
10 x 100 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction