Learn More
ERIVAN BIO LLC Human Cord Blood Exosomes
Supplier: ERIVAN BIO LLC UCBP 01200103
This product consists of extracellular vesicles isolated and purified from fresh human cord blood plasma. Human cord blood plasma is obtained from consented healthy donors and verified free from HIV, HCV, HBV, and Syphilis.