Learn More
Cytiva Ultrareservoir Containers - 2 L container (includes 0-4.2 bar pressure gauge, assorted fittings, and 3-way valve); suitable for use with Ultrasette device (1/pkg)
Supplier:  Cytiva FS005X75
Encompass_Preferred
Simplify Processing of 100 mL to 5 L Volumes When Using the Ultrasette™ Device-Self-contained siphoning units allow continuous buffer exchange-Uses less liquid with less handling and mess than conventional dialysis systems-Constructed of clear acrylic with easy-to-read volume graduations-Connections located in base plate for easy access and minimal sample hold-up-Feed pressure gauge monitors sample processing-Three-way sampling valve allows maximum sample recovery without decanting-Peristaltic pump action minimizes shear of sensitive molecules, eliminates air in the system, and reduces foaming-A variable speed controller allows precise adjustment of crossflow during sample processing This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3459