Learn More
DuPont™ Tychem™ 6000 Sleeve
Click to view available options
UNIVERSAL
Description
Tychem™ 6000 sleeves are made of a proprietary barrier film laminated to a Tyvek™ substrate. Tychem™ 6000 fabrics are tested by a third party laboratory to assess its chemical permeation resistance.
- Provides protection against over 180 chemical challenges for at least 30 minutes
- Used by military personnel and first responders for chemical warfare situations
- Available in low-visibility gray fabric for discretionary purpose
Certifications
Chemical Protective Clothing, Cat.III,Type Partial Body
Specifications
Material | Tychem |
Certifications/Compliance | Chemical Protective Clothing, Cat.III,Type Partial Body |