Learn More
Rockland Immunochemicals Turkey Blood in Alsevers 50 mL
SDP
Supplier:  Rockland Immunochemicals R3130050
Encompass_Preferred
Turkey whole blood is aseptically collected and prepared to order from our colony of animals. These cell suspensions are useful for the titration of complement adsorption procedures testing for agglutinins/HA assays and for the preparation of stroma as particulate reagents. Turkey red blood cells are perishable and are collected and processed upon receipt of your order.
Catalog No. 50-194-3984