Learn More
Hach Company Stablcal Reference Suspension II, 6 NTU, 100 mL
Supplier: Hach Company 2896842
Stabilised Formazine reference suspension for the determination of lucidity/opalescence according to European Pharmacopeia 6.0.