Learn More
Bel-Art™ SP Scienceware™ Polyethylene Utility Bags
Reusable, transparent bags are well-suited for shipping or storage and are especially useful as sample bags
Supplier: Bel-Art™ H131761218/EMD
Description
- FDA approved material; Latex free
- Different sizes available to best match particular packaging requirements
- Not autoclavable
Specifications
Polyethylene | |
18 x 12 in. (457 x 305mm) | |
457 mm | |
305 mm |
0.038 mm | |
18 in. | |
12 in. |